Assay Details
| Gene Symbol: fcn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006717.2 | NP_001006718.1 | 4 - 5 | 399 | 82 | |
| XM_012969759.2 | XP_012825213.1 | 3 - 4 | 345 | 82 | ||
| GenBank mRNA | BC075449.1 | - | 4 - 5 | 401 | 82 | |
More Information
Set Membership: |
|

fcn3
Genomic Map