Assay Details
| Gene Symbol: rnf126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006734.1 | NP_001006735.1 | 6 - 7 | 681 | 89 | |
| XM_012959798.2 | XP_012815252.1 | 5 - 6 | 625 | 89 | ||
| GenBank mRNA | BC075492.1 | - | 6 - 7 | 681 | 89 | |
| CR848559.2 | - | 6 - 7 | 679 | 89 | ||
More Information
Set Membership: |
|

rnf126
Genomic Map