Assay Details
| Gene Symbol: wdr34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006745.1 | NP_001006746.1 | 3 - 4 | 396 | 75 | |
| XM_012968224.2 | XP_012823678.1 | 2 - 3 | 566 | 75 | ||
| XM_012968225.2 | XP_012823679.1 | 2 - 3 | 358 | 75 | ||
| GenBank mRNA | BC075512.1 | - | 3 - 4 | 396 | 75 | |
More Information
Set Membership: |
|

wdr34
Genomic Map