Assay Details
| Gene Symbol: arhgdib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006837.1 | NP_001006838.1 | 3 - 4 | 332 | 80 | |
| GenBank mRNA | BC076917.1 | - | 3 - 4 | 332 | 80 | |
More Information
Set Membership: |
|

arhgdib
Genomic Map