Assay Details
Gene Symbol: prtn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006846.1 | NP_001006847.1 | 2 - 3 | 215 | 74 | |
GenBank mRNA | BC076933.1 | - | 2 - 3 | 215 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
