Assay Details
Gene Symbol: tra2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006877.1 | NP_001006878.1 | 7 - 8 | 890 | 66 | |
XM_012970311.2 | XP_012825765.1 | 8 - 9 | 1124 | 66 | ||
GenBank mRNA | BC077018.1 | - | 7 - 8 | 890 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
