Assay Details
Gene Symbol: rbm38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006915.1 | NP_001006916.1 | 2 - 3 | 615 | 58 | |
GenBank mRNA | BC075431.1 | - | 2 - 3 | 615 | 58 | |
CR848225.2 | - | 2 - 3 | 616 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
