Assay Details
| Gene Symbol: tmem248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007493.1 | NP_001007494.1 | 4 - 5 | 918 | 75 | |
| GenBank mRNA | BC079930.1 | - | 4 - 5 | 918 | 75 | |
| CR762290.2 | - | 4 - 5 | 875 | 75 | ||
More Information
Set Membership: |
|

tmem248
Genomic Map