Assay Details
| Gene Symbol: dleu7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007981.1 | NP_001007982.1 | 1 - 2 | 468 | 62 | |
| GenBank mRNA | BC080492.1 | - | 1 - 2 | 468 | 62 | |
| CT030469.1 | - | 1 - 2 | 453 | 62 | ||
| JQ838068.1 | - | 1 - 2 | 435 | 62 | ||
More Information
Set Membership: |
|

dleu7
Genomic Map