Assay Details
| Gene Symbol: ppif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257100.1 | NP_001244029.1 | 4 - 5 | 425 | 85 | |
| NR_046434.1 | 3 - 4 | 394 | 85 | |||
| XM_012966468.2 | XP_012821922.1 | 3 - 4 | 404 | 85 | ||
| GenBank mRNA | AL868523.2 | - | 4 - 5 | 413 | 85 | |
| BC080913.1 | - | 3 - 4 | 394 | 85 | ||
| CU075538.1 | - | 4 - 5 | 386 | 85 | ||
More Information
Set Membership: |
|

ppif
Genomic Map