Assay Details
| Gene Symbol: hbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008148.2 | NP_001008149.2 | 5 - 6 | 631 | 76 | |
| XM_012958894.2 | XP_012814348.1 | 4 - 5 | 671 | 76 | ||
| XM_012958896.2 | XP_012814350.1 | 4 - 5 | 680 | 76 | ||
| XM_018092050.1 | XP_017947539.1 | 4 - 5 | 673 | 76 | ||
| XM_018092051.1 | XP_017947540.1 | 4 - 5 | 570 | 76 | ||
| GenBank mRNA | BC081362.1 | - | 5 - 6 | 632 | 76 | |
More Information
Set Membership: |
|

hbp1
Genomic Map