Assay Details
| Gene Symbol: tmem45b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011108.1 | NP_001011108.1 | 4 - 5 | 704 | 68 | |
| XM_018095363.1 | XP_017950852.1 | 4 - 5 | 749 | 68 | ||
| GenBank mRNA | BC084505.1 | - | 4 - 5 | 704 | 68 | |
| CR760159.2 | - | 4 - 5 | 629 | 68 | ||
More Information
Set Membership: |
|

tmem45b
Genomic Map