Assay Details
| Gene Symbol: epcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011149.1 | NP_001011149.1 | 7 - 8 | 1128 | 72 | |
| GenBank mRNA | BC084898.1 | - | 7 - 8 | 1128 | 72 | |
More Information
Set Membership: |
|

epcam
Genomic Map