Assay Details
| Gene Symbol: sh3bp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011179.1 | NP_001011179.1 | 1 - 2 | 2579 | 83 | |
| GenBank mRNA | BC086304.1 | - | 1 - 2 | 2579 | 83 | |
| CT030498.1 | - | 1 - 2 | 2593 | 83 | ||
More Information
Set Membership: |
|

sh3bp4
Genomic Map