Assay Details
| Gene Symbol: fcn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011233.1 | NP_001011233.1 | 4 - 5 | 283 | 81 | |
| GenBank mRNA | BC087801.1 | - | 4 - 5 | 283 | 81 | |
More Information
Set Membership: |
|

fcn3
Genomic Map