Assay Details
| Gene Symbol: zdhhc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011290.1 | NP_001011290.1 | 4 - 5 | 683 | 85 | |
| XM_012964606.2 | XP_012820060.1 | 4 - 5 | 638 | 85 | ||
| XM_012964607.2 | XP_012820061.1 | 4 - 5 | 745 | 85 | ||
| XM_012964608.2 | XP_012820062.1 | 4 - 5 | 819 | 85 | ||
| XM_012964609.2 | XP_012820063.1 | 4 - 5 | 733 | 85 | ||
| XM_012964610.2 | XP_012820064.1 | 4 - 5 | 819 | 85 | ||
| GenBank mRNA | BC088007.1 | - | 4 - 5 | 683 | 85 | |
| CR848357.2 | - | 4 - 5 | 622 | 85 | ||
More Information
Set Membership: |
|

zdhhc3
Genomic Map