Assay Details
| Gene Symbol: hif1an | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203584.1 | NP_988915.1 | 7 - 8 | 1030 | 66 | |
| GenBank mRNA | BC061609.1 | - | 7 - 8 | 1030 | 66 | |
More Information
Set Membership: |
|

hif1an
Genomic Map