Assay Details
| Gene Symbol: krt19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203587.1 | NP_988918.1 | 6 - 7 | 1290 | 91 | |
| GenBank mRNA | BC061624.1 | - | 6 - 7 | 1290 | 91 | |
| CR761861.2 | - | 6 - 7 | 1254 | 91 | ||
More Information
Set Membership: |
|

krt19
Genomic Map