Assay Details
| Gene Symbol: agmo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011313.1 | NP_001011313.1 | 10 - 11 | 1262 | 72 | |
| GenBank mRNA | BC088041.1 | - | 10 - 11 | 1262 | 72 | |
More Information
Set Membership: |
|

agmo
Genomic Map