Assay Details
| Gene Symbol: limd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011377.1 | NP_001011377.1 | 3 - 4 | 168 | 93 | |
| XM_012971717.2 | XP_012827171.1 | 3 - 4 | 272 | 93 | ||
| GenBank mRNA | BC088558.1 | - | 3 - 4 | 168 | 93 | |
More Information
Set Membership: |
|

limd2
Genomic Map