Assay Details
| Gene Symbol: zcchc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011465.1 | NP_001011465.1 | 4 - 5 | 646 | 67 | |
| GenBank mRNA | BC088591.1 | - | 4 - 5 | 646 | 67 | |
More Information
Set Membership: |
|

zcchc9
Genomic Map