Assay Details
Gene Symbol: adh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011502.1 | NP_001011502.1 | 7 - 8 | 973 | 58 | |
GenBank mRNA | BC088898.1 | - | 7 - 8 | 973 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
