Assay Details
| Gene Symbol: fut9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015689.1 | NP_001015689.1 | 1 | 952 | 78 | |
| GenBank mRNA | AJ879583.1 | - | 1 | 952 | 78 | |
| BC121558.1 | - | 1 | 953 | 78 | ||
More Information
Set Membership: |
|

fut9
Genomic Map