Assay Details
| Gene Symbol: mospd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015761.1 | NP_001015761.1 | 3 - 4 | 376 | 61 | |
| XM_012968327.2 | XP_012823781.1 | 4 - 5 | 582 | 61 | ||
| XM_012968329.2 | XP_012823783.1 | 4 - 5 | 529 | 61 | ||
| GenBank mRNA | BC089697.1 | - | 3 - 4 | 376 | 61 | |
| BC121274.1 | - | 2 - 3 | 351 | 61 | ||
| BC171186.1 | - | 3 - 4 | 367 | 61 | ||
| BC171188.1 | - | 3 - 4 | 367 | 61 | ||
| CR848211.2 | - | 3 - 4 | 377 | 61 | ||
More Information
Set Membership: |
|

mospd1
Genomic Map