Assay Details
| Gene Symbol: dnal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016312.2 | NP_001016312.1 | 6 - 7 | 417 | 88 | |
| XM_012968391.2 | XP_012823845.1 | 5 - 6 | 660 | 88 | ||
| GenBank mRNA | BC158985.1 | - | 5 - 6 | 474 | 88 | |
| BC161581.1 | - | 6 - 7 | 431 | 88 | ||
| BC170910.1 | - | 6 - 7 | 415 | 88 | ||
| BC170912.1 | - | 6 - 7 | 415 | 88 | ||
| CR761486.2 | - | 6 - 7 | 417 | 88 | ||
More Information
Set Membership: |
|

dnal1
Genomic Map