Assay Details
| Gene Symbol: c15orf41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016489.2 | NP_001016489.1 | 10 - 11 | 688 | 100 | |
| XM_012968415.2 | XP_012823869.1 | 10 - 11 | 727 | 100 | ||
| XM_012968416.2 | XP_012823870.1 | 10 - 11 | 697 | 100 | ||
| XM_012968417.2 | XP_012823871.1 | 10 - 11 | 682 | 100 | ||
| XM_018096257.1 | XP_017951746.1 | 10 - 11 | 729 | 100 | ||
| GenBank mRNA | BC135523.1 | - | 10 - 11 | 700 | 100 | |
| CR760834.2 | - | 10 - 11 | 688 | 100 | ||
More Information
Set Membership: |
|

c15orf41
Genomic Map