Assay Details
| Gene Symbol: c15orf41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016489.2 | NP_001016489.1 | 6 - 7 | 431 | 89 | |
| XM_012968415.2 | XP_012823869.1 | 6 - 7 | 470 | 89 | ||
| XM_012968416.2 | XP_012823870.1 | 6 - 7 | 440 | 89 | ||
| XM_012968417.2 | XP_012823871.1 | 6 - 7 | 425 | 89 | ||
| XM_018096257.1 | XP_017951746.1 | 6 - 7 | 472 | 89 | ||
| GenBank mRNA | BC135523.1 | - | 6 - 7 | 443 | 89 | |
| CR760834.2 | - | 6 - 7 | 431 | 89 | ||
More Information
Set Membership: |
|

c15orf41
Genomic Map