Assay Details
| Gene Symbol: hsd17b10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016511.1 | NP_001016511.1 | 3 - 4 | 385 | 74 | |
| GenBank mRNA | BC135492.1 | - | 3 - 4 | 353 | 74 | |
| BC157293.1 | - | 3 - 4 | 355 | 74 | ||
| CR926229.1 | - | 3 - 4 | 385 | 74 | ||
More Information
Set Membership: |
|

hsd17b10
Genomic Map