Assay Details
| Gene Symbol: dcp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016863.2 | NP_001016863.1 | 9 - 10 | 1189 | 82 | |
| GenBank mRNA | CR848647.2 | - | 9 - 10 | 1189 | 82 | |
More Information
Set Membership: |
|

dcp2
Genomic Map