Assay Details
| Gene Symbol: prickle1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016939.1 | NP_001016939.1 | 7 - 8 | 1863 | 75 | |
| XM_012959068.2 | XP_012814522.1 | 7 - 8 | 1859 | 75 | ||
| GenBank mRNA | BC167911.1 | - | 7 - 8 | 1852 | 75 | |
| CR761986.2 | - | 7 - 8 | 1863 | 75 | ||
More Information
Set Membership: |
|

prickle1
Genomic Map