Assay Details
| Gene Symbol: gmfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017002.2 | NP_001017002.1 | 2 - 3 | 122 | 68 | |
| XM_018096272.1 | XP_017951761.1 | 2 - 3 | 212 | 68 | ||
| GenBank mRNA | BC135625.1 | - | 2 - 3 | 146 | 68 | |
| CR762326.2 | - | 2 - 3 | 122 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018096272.1 transcripts.
Set Membership: |
|

gmfb
Genomic Map