Assay Details
| Gene Symbol: mex3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017067.2 | NP_001017067.1 | 1 - 2 | 316 | 58 | |
| XM_012963233.2 | XP_012818687.1 | 1 - 2 | 672 | 58 | ||
| GenBank mRNA | BC161012.1 | - | 1 - 2 | 138 | 58 | |
| CR761586.2 | - | 1 - 2 | 316 | 58 | ||
More Information
Set Membership: |
|

mex3c
Genomic Map