Assay Details
| Gene Symbol: mief1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030527.1 | NP_001025698.1 | 4 - 5 | 896 | 67 | |
| GenBank mRNA | BC095913.1 | - | 4 - 5 | 896 | 67 | |
More Information
Set Membership: |
|

mief1
Genomic Map