Assay Details
Gene Symbol: crata | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032318.1 | NP_001027489.1 | 3 - 4 | 453 | 103 | |
XM_012968519.2 | XP_012823973.1 | 3 - 4 | 617 | 103 | ||
GenBank mRNA | BC096389.1 | - | 3 - 4 | 453 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
