Assay Details
| Gene Symbol: crata | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032318.1 | NP_001027489.1 | 6 - 7 | 846 | 67 | |
| XM_012968519.2 | XP_012823973.1 | 6 - 7 | 956 | 67 | ||
| GenBank mRNA | BC096389.1 | - | 6 - 7 | 846 | 67 | |
More Information
Set Membership: |
|

crata
Genomic Map