Assay Details
Gene Symbol: acbd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008064.2 | NP_001008065.2 | 1 - 2 | 453 | 84 | |
GenBank mRNA | BC080953.1 | - | 1 - 2 | 500 | 84 | |
CR848500.2 | - | 1 - 2 | 453 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
