Assay Details
| Gene Symbol: cnep1r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011315.2 | NP_001011315.1 | 97 | 72 | ||
| GenBank mRNA | BC088044.1 | - | 154 | 72 | ||
| CR760569.2 | - | 97 | 72 | |||
More Information
Set Membership: |
|

cnep1r1
Genomic Map