Assay Details
| Gene Symbol: fam3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015990.2 | NP_001015990.1 | 3 - 4 | 336 | 70 | |
| GenBank mRNA | BC154872.1 | - | 3 - 4 | 353 | 70 | |
| CR848320.2 | - | 3 - 4 | 336 | 70 | ||
More Information
Set Membership: |
|

fam3b
Genomic Map