Assay Details
| Gene Symbol: tmem150b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015997.2 | NP_001015997.1 | 5 - 6 | 402 | 69 | |
| XM_012966557.2 | XP_012822011.1 | 5 - 6 | 966 | 69 | ||
| XM_018095388.1 | XP_017950877.1 | 4 - 5 | 704 | 69 | ||
| GenBank mRNA | BC155023.1 | - | 5 - 6 | 392 | 69 | |
| CR848292.2 | - | 5 - 6 | 402 | 69 | ||
More Information
Set Membership: |
|

tmem150b
Genomic Map