Assay Details
| Gene Symbol: srek1ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015999.2 | NP_001015999.1 | 3 - 4 | 271 | 102 | |
| GenBank mRNA | BC135675.1 | - | 3 - 4 | 235 | 102 | |
| CR848286.2 | - | 3 - 4 | 271 | 102 | ||
More Information
Set Membership: |
|

srek1ip1
Genomic Map