Assay Details
| Gene Symbol: tmem104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016025.2 | NP_001016025.1 | 9 - 10 | 1008 | 61 | |
| XM_012971755.2 | XP_012827209.1 | 9 - 10 | 1008 | 61 | ||
| XM_012971756.2 | XP_012827210.1 | 9 - 10 | 1026 | 61 | ||
| GenBank mRNA | BC170737.1 | - | 9 - 10 | 805 | 61 | |
| BC170739.1 | - | 9 - 10 | 805 | 61 | ||
| CR848152.2 | - | 9 - 10 | 1008 | 61 | ||
More Information
Set Membership: |
|

tmem104
Genomic Map