Assay Details
| Gene Symbol: znf300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016118.2 | NP_001016118.1 | 442 | 87 | ||
| GenBank mRNA | CR762092.2 | - | 442 | 87 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

znf300| Gene Symbol: znf300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016118.2 | NP_001016118.1 | 442 | 87 | ||
| GenBank mRNA | CR762092.2 | - | 442 | 87 | ||