Assay Details
| Gene Symbol: cyp26a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016147.2 | NP_001016147.1 | 2 - 3 | 445 | 60 | |
| XM_012966562.2 | XP_012822016.1 | 2 - 3 | 503 | 60 | ||
| GenBank mRNA | BC167271.1 | - | 2 - 3 | 460 | 60 | |
| BC171087.1 | - | 2 - 3 | 437 | 60 | ||
| CR761993.2 | - | 2 - 3 | 445 | 60 | ||
More Information
Set Membership: |
|

cyp26a1
Genomic Map