Assay Details
| Gene Symbol: chmp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016229.2 | NP_001016229.2 | 4 - 5 | 443 | 86 | |
| XM_012962101.2 | XP_012817555.1 | 5 - 6 | 2720 | 86 | ||
| GenBank mRNA | BC091022.1 | - | 4 - 5 | 443 | 86 | |
| CR926287.1 | - | 4 - 5 | 429 | 86 | ||
More Information
Set Membership: |
|

chmp3
Genomic Map