Assay Details
Gene Symbol: sdr39u1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016261.2 | NP_001016261.1 | 2 - 3 | 171 | 61 | |
GenBank mRNA | BC157797.1 | - | 4 - 5 | 911 | 61 | |
BC170744.1 | - | 2 - 3 | 148 | 61 | ||
BC170746.1 | - | 2 - 3 | 148 | 61 | ||
CR761700.2 | - | 2 - 3 | 171 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
