Assay Details
| Gene Symbol: rfc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016363.2 | NP_001016363.1 | 1 - 2 | 154 | 63 | |
| GenBank mRNA | BC080504.1 | - | 2 - 3 | 213 | 63 | |
| BC135432.1 | - | 2 - 3 | 180 | 63 | ||
| CR761294.2 | - | 1 - 2 | 154 | 63 | ||
More Information
Set Membership: |
|

rfc4
Genomic Map