Assay Details
| Gene Symbol: cept1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016434.3 | NP_001016434.1 | 3 - 4 | 683 | 80 | |
| XM_012962314.2 | XP_012817768.1 | 5 - 6 | 982 | 80 | ||
| XM_012962318.2 | XP_012817772.1 | 4 - 5 | 877 | 80 | ||
| XM_012962328.2 | XP_012817782.1 | 5 - 6 | 982 | 80 | ||
| XM_018091250.1 | XP_017946739.1 | 5 - 6 | 982 | 80 | ||
| XM_018091251.1 | XP_017946740.1 | 4 - 5 | 914 | 80 | ||
| XM_018091252.1 | XP_017946741.1 | 4 - 5 | 879 | 80 | ||
| XM_018091253.1 | XP_017946742.1 | 4 - 5 | 773 | 80 | ||
| XM_018091254.1 | XP_017946743.1 | 4 - 5 | 740 | 80 | ||
| XM_018091255.1 | XP_017946744.1 | 5 - 6 | 982 | 80 | ||
| GenBank mRNA | BC161029.1 | - | 4 - 5 | 716 | 80 | |
| CR761040.2 | - | 3 - 4 | 684 | 80 | ||
More Information
Set Membership: |
|

cept1
Genomic Map