Assay Details
| Gene Symbol: tmed5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016560.2 | NP_001016560.1 | 1 - 2 | 272 | 89 | |
| GenBank mRNA | CR760694.2 | - | 1 - 2 | 272 | 89 | |
More Information
Set Membership: |
|

tmed5
Genomic Map