Assay Details
| Gene Symbol: fam221a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016728.2 | NP_001016728.1 | 3 - 4 | 431 | 67 | |
| XM_012964717.2 | XP_012820171.1 | 3 - 4 | 1395 | 67 | ||
| XM_012964718.2 | XP_012820172.1 | 3 - 4 | 469 | 67 | ||
| XM_012964719.2 | XP_012820173.1 | 3 - 4 | 487 | 67 | ||
| XM_012964720.2 | XP_012820174.1 | 3 - 4 | 384 | 67 | ||
| XM_018094657.1 | XP_017950146.1 | 2 - 3 | 348 | 67 | ||
| GenBank mRNA | BC121522.1 | - | 3 - 4 | 428 | 67 | |
| BC158501.1 | - | 3 - 4 | 489 | 67 | ||
| CR760043.2 | - | 3 - 4 | 431 | 67 | ||
More Information
Set Membership: |
|

fam221a
Genomic Map