Assay Details
Gene Symbol: mrps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016765.2 | NP_001016765.1 | 184 | 109 | ||
GenBank mRNA | CR855750.2 | - | 184 | 109 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
